Corning™ 96-Well Solid Black or White Polystyrene Microplates Edge
Edge
Manufactured from medical grade polystyrene for strength. Corning™ 96-Well Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
Supplier:  Corning™ 3915
 View more versions of this product
Catalog No. 07-200-590
Edge
Explore available promotions
Description
Description
  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • Specifications
    Specifications
    Black
    96
    Non-sterile
    Flat Bottom (F-well)
    Polystyrene
    75 to 200 μL
    Non-treated
    Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.