missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Costar™ Clear Multiple Well Plates
Untreated surface is hydrophobic in nature and binds bio-olecules through passive interactions. Corning™ Costar™ Clear Multiple Well Plates provide an ideal solution for immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface.
$1216.62 - $1490.17
Specifications
Format | Standard |
---|---|
Lid | With Lid |
Binding Type | Medium Binding |
Color | Clear |
Material | Polystyrene |
Catalog Number | Mfr. No. | Culture Area | Format | Volume (Metric) Well | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Culture Area | Format | Volume (Metric) Well | Price | Quantity | |||
07-201-589
|
Corning™
3737DUP |
3.8 cm2 | Standard | 6.9 mL |
Case of 100 Each for $1,406.69
|
|
|||
07-201-590
|
Corning™
3738 |
1.9 cm2 | Standard | 3.4 mL |
Case of 100 Each for $1,490.17
|
|
|||
07-201-588
|
Corning™
3736 |
9.5 cm2 | Standard | 16.8 mL |
Case of 100 Each for $1,216.62
|
|
|||
Description
- Individual alphanumerical codes for well identification, flat bottoms
- Treated for optimal cell attachment (except where noted)
- Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
- Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
- Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
- Sterile
- Nonpyrogenic
Specifications
Standard | |
Medium Binding | |
Polystyrene | |
Non-treated | |
100/Cs. |
With Lid | |
Clear | |
Sterile | |
Flat Bottom (F-well) | |
5/Pk. |