Corning™ Clear Polystyrene 96-Well Microplates
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ Clear Polystyrene 96-Well Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 96-well microplates for laboratory miniaturization and automation.
Supplier:  Corning™ 3798
 View more versions of this product
Catalog No. 07-200-106
Explore available promotions
Description
Description
  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
Specifications
Standard
8 x 12
Not Treated
EIA/RIA
96
Non-sterile
Round
100/Cs.
100/Cs.
Without Lid
4 inner packs of 25
Clear
Polystyrene
75 to 200 μL
Non-treated
Processed to improve hydrophilicity for hemagglutination and similar asays. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
25/Pk.