missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Costar™ Sterile Disposable Reagent Reservoirs

Manufactured from modified polystyrene for strength. Corning™ Costar™ Sterile Disposable Reagent Reservoirs are ideal for repetitive filling of multichannel pipetters.

Supplier:  Corning™ C4873

 View more versions of this product

Catalog No. 07-200-130


Explore available promotions

Description

Description

Manufactured from modified polystyrene for strength. Corning™ Costar™ Sterile Disposable Reagent Reservoirs are ideal for repetitive filling of multichannel pipetters.

Costar™ reagent reservoirs are ideal for repetitively filling multichannel pipettors.

  • Manufactured from modified polystyrene (white)
  • Individually wrapped, sterile
  • Disposable
Specifications

Specifications

100 mL
Polystyrene
35 mm
100/Cs.
Individually Wrapped
Single-use
145 x 85 x 35 mm
Sterile
145 mm
1/Pk.
88.5 mm
Videos
SDS
Documents

Documents

Promotions

Promotions