Corning™ Costar™ Transwell™-COL Collagen-Coated PTFE Membrane Insert
Transparent, collagen-treated microporous membranes promote cell attachment and spreading
$1442.82 - $1442.82
Specifications
Products 1
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
07-200-563
View Documents
Corning™
C3496
Case of 24 Each for $1,442.82
Description
Description
Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.
You can use cell culture inserts to:
  • Produce a cell culture environment that closely resembles an in vivo state
  • Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally
  • Co-culture cells with or without cell-to-cell contact
  • Design a diversity of experiments using various pore sizes, membrane types, and coatings Transparent, Collagen-coated PTFE membrane
  • Promotes cell attachment and spreading
  • Equimolar mixture of Collagen type I and III
  • Individually packaged
  • Multiple well plates included in each case
  • Sterile
Order Info
Individually packed and supplied in package with tissue culture plates with lids; sterile.
Specifications
Specifications
0.0
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'