Learn More
Coumarin 343, Laser Grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 405685000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Coumarin 343 | |
55804-65-4 | |
MFCD00051335 | |
Solubility in water: slightly soluble. | |
Authentic | |
285.299 | |
CHEBI:51941 | |
285.29 | |
19900L/(mole.cm) (lambda max.) | |
(for use as a laser dye) passes | |
500 mg |
240.0°C | |
C16H15NO4 | |
470nm (in basic ethanol) | |
KCDCNGXPPGQERR-UHFFFAOYSA-N | |
C1CC2=C3C(=C4C(=C2)C=C(C(=O)O4)C(=O)O)CCCN3C1 | |
108770 | |
409nm (in basic ethanol) | |
Pure | |
Glass bottle | |
Orange | |
Crystalline Powder |
Chemical Identifiers
55804-65-4 | |
285.299 | |
KCDCNGXPPGQERR-UHFFFAOYSA-N | |
CHEBI:51941 |
C16H15NO4 | |
MFCD00051335 | |
108770 | |
C1CC2=C3C(=C4C(=C2)C=C(C(=O)O4)C(=O)O)CCCN3C1 |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 259-824-6
TSCA : TSCA
RUO – Research Use Only