Learn More
Coumarin, 98%, Thermo Scientific Chemicals
$99.51 - $933.99
Chemical Identifiers
CAS | 91-64-5 |
---|---|
Molecular Formula | C9H6O2 |
Molecular Weight (g/mol) | 146.15 |
MDL Number | MFCD00006850 |
InChI Key | ZYGHJZDHTFUPRJ-UHFFFAOYSA-N |
Synonym | coumarin, 2h-chromen-2-one, 2h-1-benzopyran-2-one, cumarin, 1,2-benzopyrone, rattex, tonka bean camphor, coumarinic anhydride, coumarine, benzo-alpha-pyrone |
PubChem CID | 323 |
ChEBI | CHEBI:28794 |
SMILES | O=C1OC2=CC=CC=C2C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1533630
|
Thermo Scientific Chemicals
A1533630 |
250 g |
Each for $99.51
|
|
|||||
AAA153360B
|
Thermo Scientific Chemicals
A153360B |
1000 g |
Each for $277.78
|
|
|||||
AAA153360I
|
Thermo Scientific Chemicals
A153360I |
5000 g |
Each for $933.99
|
|
|||||
Description
Coumarin is an anticoagulant found in medicinal plants and used in phytomedicine for the treatment of venous diseases. This compound is found to have anti-tumor activities in several human cells lines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
91-64-5 | |
146.15 | |
ZYGHJZDHTFUPRJ-UHFFFAOYSA-N | |
323 | |
O=C1OC2=CC=CC=C2C=C1 |
C9H6O2 | |
MFCD00006850 | |
coumarin, 2h-chromen-2-one, 2h-1-benzopyran-2-one, cumarin, 1,2-benzopyrone, rattex, tonka bean camphor, coumarinic anhydride, coumarine, benzo-alpha-pyrone | |
CHEBI:28794 |
Specifications
91-64-5 | |
0.935 | |
162°C (323°F) | |
C9H6O2 | |
250 g | |
383644 | |
coumarin, 2h-chromen-2-one, 2h-1-benzopyran-2-one, cumarin, 1,2-benzopyrone, rattex, tonka bean camphor, coumarinic anhydride, coumarine, benzo-alpha-pyrone | |
ZYGHJZDHTFUPRJ-UHFFFAOYSA-N | |
2H-chromen-2-one | |
323 | |
146.15 | |
Coumarin |
68°C to 71°C | |
297°C to 299°C | |
Pleasant | |
MFCD00006850 | |
UN2811 | |
14,2562 | |
Soluble in water. | |
O=C1OC2=CC=CC=C2C=C1 | |
146.15 | |
CHEBI:28794 | |
98% |
Safety and Handling
GHS H Statement
H301-H351
Toxic if swallowed.
Suspected of causing cancer.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 202-086-7
RTECSNumber : GN4200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only