Invitrogen™ Countess™ II FL Reusable Slide Holder DFS Item
For use with Countess™ II FL Automated Cell Counter Only. Holds the Countess™ II FL Reusable Slide (A25750). Not compatible with Countess™ disposable slides.
Supplier:  Invitrogen™ AMEP4746
Catalog No. AMEP4746
Explore available promotions
Description
Description
Specifications
Specifications
Reusable Slide Holder
Cell Counting
1 unit
Countess
Countess™ Automated Cell Counter
Room Temperature
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.