StatLab™ CoverSafe Mounting Medium DFS Item
A fast-drying, low-hazard, optically clear, permanent mounting medium.
$101.44 - $101.44
Specifications
Product Type Mounting Medium
Reagent Type Mounting Solution
View More Specs
Alert:
Harmful if swallowed, in contact with skin, or inhaled.
Products 1
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
STLMMC0226 DFS Item
View Documents
StatLab™
MMC0226
100 mL
Each for $101.44
Description
Description
  • CoverSafe contains no xylene, no toluene, no dioxane, and no benzene
  • Pleasant citrus scent, making it an ideal low-hazard permanent mounting medium
  • For use in histology and cytology
Specifications
Specifications
Mounting Medium
Mounting Solution
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'