Covidien In-Room™ Sharps Disposal System SharpStar™ Containers
Engineered with controls to prevent overfilling and to limit accidental or intentional access to the container contents
$641.43 - $641.43
Specifications
Closure Type | Counter-Balanced Lid |
---|---|
Model | Sharpstar |
Stackable | Yes |
Autoclavable | Autoclavable |
Catalog Number | Mfr. No. | Capacity (English) | Color | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Color | Price | Quantity | ||||
22-557-177
|
Covidien
8507SA |
5 qt. | Translucent Red |
Case of 20 Each for $641.43
|
|
||||
Description
- Forced horizontal drop maximizes container capacity
- Anti-kickback feature prevents sharps from exiting the container
- “FULL” sign signifies that container has reached capacity
- Locking tabs on top of lid keeps hands away from contents during final closure
- Molded handles allow safe and secure transport of container
- Nestable containers save inventory space
Specifications
Counter-Balanced Lid | |
Sharpstar | |
Yes | |
Autoclavable |