missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Covidien Monoject™ Magellan ™ Safety Needle
One-handed operation needle-based safety technology
$26.97 - $50.00
Specifications
For Use With (Application) | Protects healthcare worker from accidental needlestick injuries |
---|---|
Includes | Robust safety shield |
Latex Free | Yes |
Product Type | Magellan™ Hypodermic Safety Needle |
Needle Point Style | Tri-Bevel |
Catalog Number | Mfr. No. | Needle Gauge | Length (English) Needle | Color | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Needle Gauge | Length (English) Needle | Color | Price | Quantity | |||
22-256-575
|
Covidien
8881850310 |
23 G | 1 in. | Blue |
Pack of 50 Each for $50.00
Case of 10 Pack for $500.00
|
|
|||
22-256-577
|
Covidien
8881850510 |
25 G | 1 in. | Orange |
Pack of 50 Each for $26.97
|
|
|||
Specifications
Protects healthcare worker from accidental needlestick injuries | |
Yes | |
Tri-Bevel |
Robust safety shield | |
Magellan™ Hypodermic Safety Needle | |
Sterile |