Covidien Phlebotomy Sharps Containers
Portable phlebotomy containers fit into present blood drawing trays for the efficient disposal of blood, hypodermic and IV needles
 View more versions of this product
Catalog No. 14-830-107A
Explore available promotions
Description
Description
  • Compact design adapts to blood drawing trays and techniques
  • Clear tops allow user to monitor fill levels
  • Container locks, securing contents for final disposal
  • Variety of opening sizes allows for disposal of a variety of sharps such as lancets, butterfly tubing, and small syringes
  • Available with holders and brackets to maintain upright throughout use
Specifications
Specifications
0.9 L
1 qt.
4.50 in.
4.25 in.
Autodrop phlebotomy
6.25 in.