Covidien Sharps-A-Gator™ Large Volume Sharps Container
Protects and ensures the safety of both healthcare professionals and patients
Supplier:  Covidien 31143665
Catalog No. 22-037-966
Explore available promotions
Description
Description
  • Large design allows for the convenient disposal of large instruments and high volumes
  • Versatile lid provides both a wide opening for larger instruments and an integrated sharps port for smaller items
  • Gasketed containers available for spill resistance
  • Clear lid keeps contents and fill level visible for increased safety
  • Convenient, molded carrying handles assist in lifting heavy, filled containers for final disposal
  • Container color: Red
Specifications
Specifications
37.8 L
10 gal.
Plastic
Clear
Red