missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CPC™ Plastic Quick-Disconnect Fittings DFS Item

Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables tests.

Supplier:  CPC™ MPC22004T39M

 View more versions of this product

Catalog No. 05-113-149


Explore available promotions

Description

Description

Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables

CPC Series couplings assure secure connections, and are easy to operate—even with gloved hands. They are reusable, yet economical enough to allow disposability.

  • Material: Medical-grade ABS, polycarbonate and polysulfone
  • Tubing Range: 1/4 and 3/8 in. I.D. (6.4 and 9.5mm)
  • Fow Rate: Cv∽5.5 (79.25) max.

  • Ergonomic thumb latch
  • Audible 'click' when connected
  • USP Class VI materials
  • Can be sterilized by autoclave, EtO, e-beam, or gamma
  • Coupling halves can be rotated when connected
  • Tapered barb
  • BSE/TSE free materials

  • Cell culture media packaging and transfer lines
  • Harvest and collection applications
  • Buffer solutions
  • Tube sets
  • Urology drainage/collection systems
  • Hypo/hyperthermia blankets

Specifications

Specifications

Hose Barb Non-valved Coupling Insert
Autoclavable
Polysulfone
33 mm
Cell culture media packaging and transfer lines, harvest and collection applications
-40°C
0.6 in.
Amber
300°F
USP Class VI, ADCF
6.4 mm
1.3 in.
6.4mm
-40°F
15.2 mm
Platinum-cured Silicone
60 psi
149°C
Videos
SDS
Documents

Documents

Promotions

Promotions