missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
CPC™ Quick-Disconnect Fitting, Sealing Plug, Polycarbonate, 1/4 in. Flow Size
Detach these fittings with a simple press of the thumb latch. Cole-Parmer™ CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate materials pass all physicochemical, cytotoxicity, leachables and extractables tests.
Supplier: CPC™ MPC30003MB25
Description
Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate materials pass all physicochemical, cytotoxicity, leachables and extractables
CPC Series couplings assure secure connections, and are easy to operate—even with gloved hands. They are reusable, yet economical enough to allow disposability.
- Material: Medical-grade ABS, polycarbonate and polysulfone
- Tubing Range: 1/4 and 3/8 in. I.D. (6.4 and 9.5mm)
- Flow Rate: Cvâ½5.5 (79.25) max.
- Ergonomic thumb latch
- Audible 'click' when connected
- USP Class VI materials
- Can be sterilized by autoclave, EtO, e-beam, or gamma
- Coupling halves can be rotated when connected
- Tapered barb
- BSE/TSE free materials
- O-Ring: Silicone Seal USP Class VI
- Cell culture media packaging and transfer lines
- Harvest and collection applications
- Buffer solutions
- Tube sets
- Urology drainage/collection systems
- Hypo/hyperthermia blankets
Specifications
Quick Disconnect Fitting | |
Autoclavable | |
19.1 mm | |
N/A | |
Purple |
USP Class VI, ADCF | |
Polycarbonate | |
31.5 mm | |
-40°C | |
121° C |