missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
CPC™ MPX Series Connectors and Configurations
Supplier: CPC™ MPX17803
Specifications
USP Class VI, ADCF | |
Polycarbonate | |
32.5 mm | |
-40°C | |
1.96 in. | |
60 psi | |
121°C |
Autoclavable | |
1.28 in. | |
-40°F | |
50 mm | |
Purple | |
250°F | |
Hose Barb Non-valved Coupling Body |