CPC™ Plastic Connectors, Couplings and Fittings DFS Item
Cover a wide range of general purpose applications with these plastic quick disconnect couplings. CPC™ Plastic Connectors, Couplings and Fittings feature integral terminations and one-hand connection and disconnection.
Supplier:  CPC™ PLCD22005
 View more versions of this product
Catalog No. 05-114-014
Explore available promotions
Description
Description
  • Available in 1/8 to 1/2 in. nominal flow
  • Polypropylene construction
  • EPDM 0-rings
  • Colder thumb latch
  • Chemically resistant and gamma sterilizable
  • Fewer leak points, shorter assemblies and faster installations
Specifications
Specifications
Hose Barb Valved In-line Coupling Insert
Acetal
34.2 mm
-40°F
16.5 mm
Plastic connector
120 psi
80°C
1.35 L x 0.62 in. (34.2 x 16.5 mm)
1.35 in.
8mm
-40°C
0.62 in.
White
180°F