missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Creatine phosphate, disodium salt, 99.12%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210052025

 View more versions of this product

Catalog No. ICN10052025


Explore available promotions

Description

Description

Specifications

Specifications

Creatine phosphate, disodium salt
922-32-7
25 g
C4H8N3Na2O5P
creatine phosphate sodium salt
[Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O
255.08
327.2
White
Crystalline Powder
98-100%
MFCD00150192
RNTXMYSPASRLFT-UHFFFAOYSA-L
disodium ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
131887655

Chemical Identifiers

922-32-7
255.08
RNTXMYSPASRLFT-UHFFFAOYSA-L
131887655
[Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O
C4H8N3Na2O5P
MFCD00150192
creatine phosphate sodium salt
disodium ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at - 0°C

SDS
Documents

Documents

Promotions

Promotions