missing translation for 'onlineSavingsMsg'
Learn More

Creatine phosphate disodium salt tetrahydrate, 98+%

CAS: 71519-72-7 | C4H8N3Na2O5P·4H2O

$76.79 - $979.74

Chemical Identifiers

CAS 71519-72-7
Molecular Formula C4H8N3Na2O5P·4H2O
MDL Number MFCD00150192
Synonym Sodium creatine phosphate dibasic tetrahydrate
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1536203
View Documents
Thermo Scientific Chemicals
A1536203
1 g
Each for $76.79
Only null left
 
AAA1536206
View Documents
Thermo Scientific Chemicals
A1536206
5 g
Each for $257.74
Only null left
 
AAA1536214
View Documents
Thermo Scientific Chemicals
A1536214
25 g
Each for $979.74
Only null left
 
Description

Description

Creatine phosphate disodium salt acts as a substrate for creatine kinase determinations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Creatine phosphate disodium salt acts as a substrate for creatine kinase determinations.

Solubility
Soluble in water.0.1 g/mL.

Notes
Incompatible with Strong bases. strong acids. Protect from Direct sunlight.
Specifications

Chemical Identifiers

71519-72-7
C4H8N3Na2O5P·4H2O
MFCD00150192
Sodium creatine phosphate dibasic tetrahydrate

Specifications

71519-72-7
C4H8N3Na2O5P·4H2O
1 g
327.14 (255.07 Anhydrous)
Powder
≥98%
MFCD00150192
Soluble in water.0.1g/mL.
≥98%
Creatine Phosphate Disodium Salt Tetrahydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 213-074-6

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only