missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine phosphate disodium salt tetrahydrate, 99%
CAS: 71519-72-7 | C4H8N3Na2O5P | 255.08 g/mol
$147.55 - $618.46
Chemical Identifiers
| CAS | 71519-72-7 |
|---|---|
| Molecular Formula | C4H8N3Na2O5P |
| Molecular Weight (g/mol) | 255.08 |
| MDL Number | MFCD00150192 |
| InChI Key | RNTXMYSPASRLFT-UHFFFAOYSA-L |
| Synonym | creatine phosphate sodium salt |
| PubChem CID | 91884830 |
| SMILES | [Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC337340010
|
Thermo Scientific Chemicals
337340010 |
1 g | Glass bottle |
Each for $147.55
|
|
||||
|
AC337340020
|
Thermo Scientific Chemicals
337340020 |
2 g | Glass bottle |
Each for $274.03
|
|
||||
|
AC337340100
|
Thermo Scientific Chemicals
337340100 |
10 g | Glass bottle |
Each for $618.46
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 71519-72-7 | |
| 255.08 | |
| RNTXMYSPASRLFT-UHFFFAOYSA-L | |
| 91884830 |
| C4H8N3Na2O5P | |
| MFCD00150192 | |
| creatine phosphate sodium salt | |
| [Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O |
Specifications
| 71519-72-7 | |
| Authentic | |
| Glass bottle | |
| Na2O3PNHC(NH)N(CH3)CH2COOH·4H2O | |
| 1 g | |
| (10% in water) Clear colorless solution | |
| [Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O | |
| 255.08 | |
| 327.14 | |
| Powder |
| White | |
| 99% | |
| C4H8N3Na2O5P | |
| MFCD00150192 | |
| creatine phosphate sodium salt | |
| RNTXMYSPASRLFT-UHFFFAOYSA-L | |
| disodium tetrahydrate ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate | |
| 91884830 | |
| 99% | |
| Creatine phosphate disodium salt tetrahydrate |
RUO – Research Use Only