missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Supelco™ Crimp seals with PTFE/Rubber Septa Red Aluminum Seal
Closure type crimp seal
$300.00 - $300.00
Specifications
Diameter (Metric) Inner | 11 mm |
---|---|
Color | Red |
Septum | Rubber |
For Use With (Equipment) | 11 mm Crimp Top Vial |
Catalog Number | Mfr. No. | No. per Pack | Material | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. per Pack | Material | Price | Quantity | |||||
111009837
|
MilliporeSigma™ Supelco™
27473 |
1000 | Aluminum (Seal), PTFE |
Each for $300.00
|
|
|||||
Description
- The PTFE/rubber septa are compatible with most organic solvents until pierced and are not recommended for aromatics, carbon disulfides, chlorinated solvents, hydrocarbons.
Specifications
11 mm | |
Red | |
Rubber | |
11 mm Crimp Top Vial |