missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
CRMP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Fisher Scientific PA529728
Description
Recommended positive controls: U87-MG, SK-N-SH, IMR32, SK-N-AS, mouse brain, rat brain. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (87%), Chicken (95%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, neuronal growth cone collapse and cell migration.Specifications
CRMP2 | |
Polyclonal | |
Unconjugated | |
DPYSL2 | |
AI851130; collapsin response mediator protein 2; collapsin response mediator protein hCRMP-2; CRMP2; CRMP-2; DHPRP2; dihydropyrimidinase like 2; dihydropyrimidinase-like 2; dihydropyrimidinase-related protein 2; DPYL2; Dpysl2; DRP2; DRP-2; Musunc33; N2A3; Neural-specific protein NSP60; TOAD-64; turned on after division 64; turned on after division 64 kDa protein; turned on after division, 64 kDa protein; ULIP2; ULIP-2; unc-33-like phosphoprotein 2; Unknown (protein for MGC:137406) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12934, 1808, 25416 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.7 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
O08553, P47942, Q16555 | |
DPYSL2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 219 of Human CRMP2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |