missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Croconic acid disodium salt, 97%, Thermo Scientific Chemicals
$103.55 - $255.67
Chemical Identifiers
CAS | 14379-00-1 |
---|---|
Molecular Formula | C5Na2O5 |
Molecular Weight (g/mol) | 186.03 |
MDL Number | MFCD00191954 |
InChI Key | OQXLFPHHAAAVKQ-UHFFFAOYSA-L |
Synonym | croconic acid disodium salt, croconic acid, disodium salt, disodium trioxocyclopent-1-ene-1,2-bis olate, sodium 3,4,5-trioxocyclopent-1-ene-1,2-bis olate, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione disodium salt |
PubChem CID | 12120285 |
IUPAC Name | disodium;3,4,5-trioxocyclopentene-1,2-diolate |
SMILES | [Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C1=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2223501
|
Thermo Scientific Chemicals
B2223501 |
0.25 g |
Each for $103.55
|
|
|||||
AAB2223503
|
Thermo Scientific Chemicals
B2223503 |
1 g |
Each for $255.67
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14379-00-1 | |
186.03 | |
OQXLFPHHAAAVKQ-UHFFFAOYSA-L | |
12120285 | |
[Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C1=O |
C5Na2O5 | |
MFCD00191954 | |
croconic acid disodium salt, croconic acid, disodium salt, disodium trioxocyclopent-1-ene-1,2-bis olate, sodium 3,4,5-trioxocyclopent-1-ene-1,2-bis olate, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione disodium salt | |
disodium;3,4,5-trioxocyclopentene-1,2-diolate |
Specifications
14379-00-1 | |
C5Na2O5 | |
0.25 g | |
OQXLFPHHAAAVKQ-UHFFFAOYSA-L | |
disodium;3,4,5-trioxocyclopentene-1,2-diolate | |
12120285 | |
97% |
>300°C | |
MFCD00191954 | |
croconic acid disodium salt, croconic acid, disodium salt, disodium trioxocyclopent-1-ene-1,2-bis olate, sodium 3,4,5-trioxocyclopent-1-ene-1,2-bis olate, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione disodium salt | |
[Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C1=O | |
186.03 | |
186.03 | |
Croconic acid disodium salt |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only