DWK Life Sciences Kimble™ Cross Stir Bar
This all-purpose stir bar permits stirring in closed containers without contamination. Ideal for use with conical bottom HPLC reservoirs.
$65.09 - $75.68
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity | ||||||
K7911900100
|
DWK Life Sciences
7911900100 |
Fits 2L Reservoir |
Case for $65.09
|
|
||||||
K7911900114
|
DWK Life Sciences
7911900114 |
Fits 5L, 10L, 20L Reservoirs |
Case for $75.68
|
|
||||||
Description
- PTFE-covered X-shaped magnetic stirring bar
- More turbulence with less vibration than straight stir bars
- Autoclavable