missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ad Infinitum™ CryoProtectPURE™ Cryopreservation Media
Ad Infinitum™ CryoProtectPURE™ Cryopreservation Media is a DMSO-Free and Serum-Free cryopreservation media for cell and tissue preservation.
$643.91 - $9,658.62
Specifications
Certifications/Compliance | ISO 13485 standards |
---|---|
Content And Storage | 2 to 8°C |
Description | Pre-mixed and ready-to-use |
Product Type | Cryopreservation Media |
For Use With (Application) | Manufacturing and/or research use only, not for diagnostic or therapeutic use |
Description
- Increases the cell viability during freeze and post-thaw, producing healthier cells with no antigenicity concern or toxic vitrification.
- Serum-Free
- Pre-mixed
- Ready-to-use
- 0.2 μm filtered
- Suitable for a wide variety of cells
- Shipped at ambient temperature
- For manufacturing and/or research use only, not for diagnostic or therapeutic use
- Manufactured at a cGMP-compliant facility registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards
Specifications
ISO 13485 standards | |
Pre-mixed and ready-to-use | |
Manufacturing and/or research use only, not for diagnostic or therapeutic use |
2 to 8°C | |
Cryopreservation Media |