missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Crystal Violet Indicator, 1% (w/v) in Glacial Acetic Acid, Ricca Chemical

Gentian Violet Indicator for Nonaqueous Titrations

Supplier:  Ricca Chemical Company 24834

 View more versions of this product

Catalog No. 2483-4


Explore available promotions

Description

Description

Specifications

Specifications

Crystal Violet (Gentian Violet) Indicator
1% (w/v)
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
11057
Amber Glass Bottle
Blue to Purple/Violet
Liquid
548-62-9,64-19-7
C25H30ClN3
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
407.986
CHEBI:41688
Glacial Acetic Acid
120 mL

Chemical Identifiers

548-62-9
407.986
11057
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
C25H30ClN3
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
CHEBI:41688
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Videos
SDS Documents

Documents

Promotions

Promotions