Learn More
Gibco™ CTS™ GlutaMAX™ Supplement
CTS GlutaMAX Supplement is an animal origin free (AOF) dipeptide alternative to L-glutamine, with increased stability that improves cell health.
Supplier: Gibco™ A4737001
Description
CTS GlutaMAX Supplement is an animal origin free (AOF) dipeptide alternative to L-glutamine, with increased stability that improves cell health. CTS GlutaMAX Supplement can be used as a direct substitute for L-glutamine at equimolar concentrations in mammalian and stem cell culture with minimal or no adaptation.
Gibco CTS products are designed for use in manufacturing of cell or gene products, supporting your transition from research to clinical applications.
Compared to L-glutamine, CTS GlutaMAX Supplement:
• Minimizes toxic ammonia build-up—Unlike L-glutamine, CTS GlutaMAX Supplement does not spontaneously break down to form ammonia. This prevents waste build-up and maintains a fresh supply of L-glutamine during long-term culture.
• Improves growth efficiency and performance of mammalian cell cultures—Lower levels of ammonia result in improved cell viability. In addition, culture conditions are more controlled due to the consistent concentration of CTS GlutaMAX Supplement that is not possible with L-glutamine.
• Remains stable across a wide range of temperatures—CTS GlutaMAX Supplement retains full functionality at room temperature or 2–8°C, for convenient storage and handling.
Closed systems compatible
This size of CTS GlutaMAX Supplement is provided in bioprocess container format with the chamber constructed from Aegis 5-14 film. Sterile, weldable C-Flex™ tubing with a female luer lock provides versatile connection options for integrating into a closed system manufacturing process.
CTS quality
The Gibco CTS product line enables you to reduce your burden in qualifying reagents during your transition from research to clinical applications.
Products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485- and ISO 9001–certified. Gibco CTS products follow USP 'ancillary materials for cell, gene, and tissue-engineered products' within the responsibilities applicable to a supplier.
Gibcbo CTS products are of high quality and are supplied with harmonized documentation such as certificates of analysis, certificates of origin, and convenient access to our drug master file (DMF).
Specifications
Store at 15°C to 30°C. | |
100X | |
GlutaMax | |
Sustainable packaging | |
Room Temperature |
Animal Origin-free | |
Liquid | |
Bag - BioProcess Container (BPC) | |
100 mL |