missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ CTS™ OpTmizer™ T Cell Expansion SFM, bag format
CTS™ OpTmizer™ T Cell Expansion SFM, bag format
Supplier: Gibco™ A1048503
Description
Gibco™ OpTmizer™ CTS™ T-Cell Expansion SFM has been developed for the growth and expansion of human T lymphocytes. OpTmizer™ CTS™ T-Cell Expansion medium is a complete serum-free and xeno-free 1X medium consisting of OpTmizer™ T-Cell Expansion Basal Medium (1 L bag) and OpTmizer™ T-Cell Expansion Supplement (26 mL), which are mixed together prior to use.Versatile T Cell Culture Media
OpTmizer™ CTS™ T-Cell Expansion SFM is designed to give results equivalent to conventional serum-supplemented media. However, if desired, 2% heat-inactivated human serum (Cat. No. 34005-100) may be added to the medium to enhance viability and expansion. The use of serum, and the amount required, should be determined empirically depending on the specific T-cell culture application.
Ready for T Cell Expansion
OpTmizer™ CTS™ T-Cell Expansion SFM is designed for the expansion of human T-cells permitting CD3+ densities of >3x106 cells/mL in static conditions and >2x107 cells/mL in WAVE bags. Results may vary depending on the T-cell subset (e.g., Ag-specific cells). Conditions for specific T-cell culture applications should be determined empirically.
Stability of Complete Media
Complete OpTmizer™ CTS™ T-Cell Expansion SFM (basal, supplement, and L-glutamine) is stable for 4 weeks when stored in the dark at 2–8°C.
Simplify Your Transition into Clinical Research
The Gibco™ Cell Therapy Systems (CTS™) brand offers a broad array of products for use in cell therapy applications, including media, reagents, growth factors, enzymes, cell selection/expansion reagents, and devices. Gibco™ CTS™ products deliver:
Specifications
OpTmizer™ T-Cell Expansion Basal Medium (1 x L media bag): Store at 2–8°C. Protect from light. OpTmizer™ T-Cell Expansion Supplement (1 x 26 mL): Store in the dark at 2–8°C. |
|
T Cells | |
Suspension Cell Culture | |
Cell Expansion Serum Free Medium (SFM) | |
Sterile | |
No Glutamine | |
No antibiotics | |
Research Grade |
CO2 | |
Chemically-defined, Serum-free, Xeno-free | |
Liquid | |
Serum-free | |
HEPES, Phenol Red, Sodium Pyruvate, Sodium Bicarbonate | |
Very Low | |
Calcium, Magnesium | |
1 Bag |