Sartorius Cubis™ II Analytical Balance, MCE User Interface
Cubis™ II Analytical Balance features fully customizable hardware, software, and connectivity
$13204.90 - $24331.80
Specifications
Connection Type 2 USB A ports, 1 USB B port, 1USP C port, 1 to 9 pin RS 232 connection, 1 ethernet port
Display Type Color TFT Touchscreen
Material Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
Stabilization Time 1 sec.
Warranty 10 years
View More Specs
Products 16
Catalog Number Mfr. No. Capacity Description Includes Price Quantity  
Catalog Number Mfr. No. Capacity Description Includes Price Quantity  
14561136
View Documents Promotion Details
Sartorius
MCE124S2S00A
120 g Draft shield A - Motorized Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561135
View Documents Promotion Details
Sartorius
MCE224S2S00A
220 g Draft shield A - Motorized Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561131
View Documents Promotion Details
Sartorius
MCE524S2S00A
520 g Draft shield A - Motorized Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561134
View Documents Promotion Details
Sartorius
MCE324P2S00A
80, 160, 320 g Draft shield A - Motorized Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561133
View Documents Promotion Details
Sartorius
MCE324S2S00A
320 g Draft shield A - Motorized Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561132
View Documents Promotion Details
Sartorius
MCE524P2S00A
120, 240, 520 g Draft shield A - Motorized Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561146
View Documents Promotion Details
Sartorius
MCE124S2S00I
120 g Draft shield I - Motorized with ionizer Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561145
View Documents Promotion Details
Sartorius
MCE224S2S00I
220 g Draft shield I - Motorized with ionizer Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561143
View Documents Promotion Details
Sartorius
MCE324S2S00I
320 g Draft shield I - Motorized with ionizer Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561141
View Documents Promotion Details
Sartorius
MCE524S2S00I
520 g Draft shield I - Motorized with ionizer Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561144
View Documents Promotion Details
Sartorius
MCE324P2S00I
80, 160, 320 g Draft shield I - Motorized with ionizer Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561142
View Documents Promotion Details
Sartorius
MCE524P2S00I
120, 240, 520 g Draft shield I - Motorized with ionizer Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561266
View Documents Promotion Details
Sartorius
MCE224SUS00
220 g Without Draft Shield Balance, Accessories, Power supply, Manual, Factory test report; QP99 App Package N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561124
View Documents Promotion Details
Sartorius
MCE324P2S00U
80, 160, 320 g Draft shield U - Manual glass Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561121
View Documents Promotion Details
Sartorius
MCE524S2S00U
520 g Draft shield U - Manual glass Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561122
View Documents Promotion Details
Sartorius
MCE524P2S00U
120, 240, 520 g Draft shield U - Manual glass Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor (draft shield M), or gesture sensor (optional) learning capability
Warranty
10-year warranty
Specifications
Specifications
2 USB A ports, 1 USB B port, 1USP C port, 1 to 9 pin RS 232 connection, 1 ethernet port
Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
10 years
240 mm
100/240 V ±10%
Color TFT Touchscreen
1 sec.
Analytical
120/240 V
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'