Sartorius Cubis™ II Micro Balance, MCE User Interface
Cubis™ II Micro Balance features fully customizable hardware, software, and connectivity
$40061.70 - $66386.93
Specifications
Connection Type 2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
Display Type Color TFT Touchscreen
Material Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
Type Micro Balance
Warranty 10 years
View More Specs
Products 4
Catalog Number Mfr. No. Model Capacity Includes Price Quantity  
Catalog Number Mfr. No. Model Capacity Includes Price Quantity  
14561110
View Documents Promotion Details
Sartorius
MCE106S2S00F
Draft shield F: Filtered 10.1 g Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561112
View Documents Promotion Details
Sartorius
MCE36P2S00F
Draft shield F: Filtered 1.1 g, 2.1 g, 3.1 g Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561114
View Documents Promotion Details
Sartorius
MCE106S2S00M
Draft shield M: Motorized 10.1 g Balance, Accessories, Power supply, Manual, Factory test report N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14561268
View Documents Promotion Details
Sartorius
MCE36PMS00
Draft shield M: Motorized 1.1 g, 2.1 g, 3.1 g Balance, Accessories, Power supply, Manual, Factory test report, QP99 software N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor (draft shield M), or gesture sensor (optional) learning capability
Warranty
10-year warranty
Specifications
Specifications
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
10 years
50/60 Hz
Color TFT Touchscreen
Micro Balance
100/240 V ±10%
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'