Sartorius Cubis II MTS Balance
The combinability of the display unit, weighing module, draft shield, software packages for various applications and functions and a comprehensive range of accessories allows the individual adaptation of the Cubis™ II balance to all weighing tasks.
$11658.48 - $23788.15
Specifications
Connection Type D-sub connector, 9-pin
Display Type Color Touchscreen
Model Cubis II MTS Balance
Type Balance
Electrical Requirements 00 to 240 VAC (±10%)
View More Specs
Products 3
Catalog Number Mfr. No. Capacity Readability Description Price Quantity  
Catalog Number Mfr. No. Capacity Readability Description Price Quantity  
01-288-034
View Documents
Sartorius
MCE8202S0MTS
8200 g 10 mg MCE Display with factory-installed essential weighing applications N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-288-035
View Documents
Sartorius
MCA8202S0MTS
8200 g 10 mg MCA Display including touch, scroll, swipe functionality with factory-installed essential weighing applications and diverse QPapp Packages for optional software extensions N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-288-036
View Documents
Sartorius
MCA125SUMTS
120 g 0.01 MCA Display including touch, scroll, swipe functionality with factory-installed essential weighing applications and diverse QPapp Packages for optional software extensions N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Leading performance.
  • Monolithical weighing system, integrated climate sensors, individual sample holders
  • Error-free operation.
  • Individual QApp workflows, motorized auto-leveling
  • The modular weighing system Cubis™ II can be individually configured from different components.
Specifications
Specifications
D-sub connector, 9-pin
Cubis II MTS Balance
00 to 240 VAC (±10%)
Color Touchscreen
Balance
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'