missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ ExpiCHO™ Expression Medium

A GMP-manufactured, chemically-defined, serum-free, protein-free, animal origin–free medium specifically designed for high-density growth and transfection of suspension-adapted Chinese hamster ovary (CHO) cells.

Supplier:  Gibco™ A2910001

 View more versions of this product

Catalog No. A2910001


Explore available promotions

Description

Description

ExpiCHO Expression Medium is specifically designed for high-density growth and transfection of suspension-adapted Chinese hamster ovary (CHO) cells. This medium is GMP-manufactured, chemically-defined, serum-free, protein-free, and animal origin–free. It is a core component of the ExpiCHO Expression System Kit (Cat. No. A29133) for scalable transient protein production. To achieve the highest levels of transient protein expression, the medium is matched to a culture feed, ExpiCHO Feed, also included in the system kit and transfection kits.

With ExpiCHO Expression Medium you can:

  • Grow suspension CHO cultures to densities of over 2 x 107 cells per mL, with >95% cell viability
  • Achieve high transfection efficiencies of >90% when coupled with ExpiFectamine CHO transfection reagent
  • Transiently transfect suspension CHO cells at a density of 6 x 106 cells per mL
  • Achieve yields up to 3 g per L of recombinant protein of high-density transiently transfected cultures by sustained, high-level expression
  • Grow and transfect CHO cells at volumes of <1 mL up to 5 L in culture formats from multiwell plates up to 5 L shake flask

ExpiCHO Expression Medium is ready to use. It is formulated with GlutaMAX-I reagent and does not require supplementation. It does not contain phenol red. The chemically-defined, animal origin free formulation results in high reproducibility and lot-to-lot reliability. ExpiCHO Expression Medium is not recommended for adherent cell cultures.

Specifications

Specifications

Storage conditions: in the dark at 2-8°C
Shipping conditions: ambient
Shelf life: 12 months from date of manufacture
Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Liquid
Serum-free
No Phenol Red
ExpiCHO™
Room Temperature
ExpiCHO-S™, CHO-S, SFM Adapted
Suspension Cell Culture
CHO Expression Medium
GlutaMAX
Bottle
1000 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions