Learn More
Gibco™ ExpiCHO™ Expression Medium
A GMP-manufactured, chemically-defined, serum-free, protein-free, animal origin–free medium specifically designed for high-density growth and transfection of suspension-adapted Chinese hamster ovary (CHO) cells.
Supplier: Gibco™ A2910002
Description
ExpiCHO Expression Medium is specifically designed for high-density growth and transfection of suspension-adapted Chinese hamster ovary (CHO) cells. This medium is GMP-manufactured, chemically-defined, serum-free, protein-free, and animal origin–free. It is a core component of the ExpiCHO Expression System Kit (Cat. No. A29133) for scalable transient protein production. To achieve the highest levels of transient protein expression, the medium is matched to a culture feed, ExpiCHO Feed, also included in the system kit and transfection kits.
With ExpiCHO Expression Medium you can:
- Grow suspension CHO cultures to densities of over 2 x 107 cells per mL, with >95% cell viability
- Achieve high transfection efficiencies of >90% when coupled with ExpiFectamine CHO transfection reagent
- Transiently transfect suspension CHO cells at a density of 6 x 106 cells per mL
- Achieve yields up to 3 g per L of recombinant protein of high-density transiently transfected cultures by sustained, high-level expression
- Grow and transfect CHO cells at volumes of <1 mL up to 5 L in culture formats from multiwell plates up to 5 L shake flask
ExpiCHO Expression Medium is ready to use. It is formulated with GlutaMAX-I reagent and does not require supplementation. It does not contain phenol red. The chemically-defined, animal origin free formulation results in high reproducibility and lot-to-lot reliability. ExpiCHO Expression Medium is not recommended for adherent cell cultures.
Specifications
Storage conditions: in the dark at 2-8°C Shipping conditions: ambient Shelf life: 12 months from date of manufacture |
|
Animal Origin-free, Chemically-defined, Protein-free, Serum-free | |
Liquid | |
Serum-free | |
No Phenol Red | |
ExpiCHO™ | |
Room Temperature |
ExpiCHO-S™, CHO-S, SFM Adapted | |
Suspension Cell Culture | |
CHO Expression Medium | |
GlutaMAX | |
Bottle | |
6 x 1 L |