missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ PYREX™ Culture Tubes

All sizes except 6, 22, and 25mm O.D. have marking spot

Supplier:  Corning™ 982016EMD

 View more versions of this product

Catalog No. S63297



Explore available promotions

Description

Description

  • Rimless tops for greater convenience in plugging and rack storage
  • Reusable with walls and bottoms with uniform thickness
  • Ends are fire polished
Specifications

Specifications

Fire Polished
100 mm
Reusable
White Marking Spot
Non-sterile
16 mm
11 mL
Borosilicate Glass
For convenient plugging and rack storage
No
No
Videos
SDS
Documents

Documents

Promotions

Promotions