missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cumene, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A118640F

 View more versions of this product

Catalog No. AAA118640F


Explore available promotions

Description

Description

Cumene is used in the production of phenol and acetone. It is used as a thinner for paints, lacquers, and enamels and as a component of high octane fuels. It is used in the manufacture of phenol, acetone, acetophenone, and methylstyrene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cumene is used in the production of phenol and acetone. It is used as a thinner for paints, lacquers, and enamels and as a component of high octane fuels. It is used in the manufacture of phenol, acetone, acetophenone, and methylstyrene.

Solubility
Soluble in alcohol, ether, acetone, benzene, carbon tetrachloride. Insoluble in water.

Notes
Store at 2-8°C. Avoid prolonged storage with exposure to air. Incompatible with acids, oxidizing materials.
Specifications

Specifications

Cumene
-96°C
152°C to 154°C
99%
1.491
2500 mL
1236613
isopropylbenzene, 2-phenylpropane, cumol, benzene, 1-methylethyl, 1-methylethyl benzene, isopropylbenzol, cumeen, isopropilbenzene, isopropylbenzeen, 2-fenilpropano
RWGFKTVRMDUZSP-UHFFFAOYSA-N
cumene
7406
120.2
Liquid
98-82-8
0.864
46°C (114°F)
C9H12
MFCD00008881
UN1918
14,2617
Soluble in alcohol,ether,acetone,benzene,carbon tetrachloride. Insoluble in water.
CC(C)C1=CC=CC=C1
120.195
CHEBI:34656
99%

Chemical Identifiers

98-82-8
120.195
RWGFKTVRMDUZSP-UHFFFAOYSA-N
7406
cumene
C9H12
MFCD00008881
isopropylbenzene, 2-phenylpropane, cumol, benzene, 1-methylethyl, 1-methylethyl benzene, isopropylbenzol, cumeen, isopropilbenzene, isopropylbenzeen, 2-fenilpropano
CHEBI:34656
CC(C)C1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H304-H226-H302-H335
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Harmful if swallowed.
May cause respiratory irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P271-P281-P301+P310-P303+P361+P353-P304+P340-P308+P313-P331-P370+P378q-P501c

H226-H304-H335-H350

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ISOPROPYLBENZENE

EINECSNumber : 202-704-5

RTECSNumber : GR8575000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only