missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cupric Oxide, Black, Powdered, ACS, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor C1415500GM
Specifications
1317-38-0 | |
500 g | |
MFCD00010979 | |
[O--].[Cu++] | |
79.55 | |
ACS |
1 | |
CuO | |
KKCXRELNMOYFLS-UHFFFAOYSA-N | |
copper(2+) oxidandiide | |
99% | |
Amber Glass Bottle |
Chemical Identifiers
1317-38-0 | |
79.55 | |
KKCXRELNMOYFLS-UHFFFAOYSA-N | |
[O--].[Cu++] |
CuO | |
MFCD00010979 | |
copper(2+) oxidandiide |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.