Learn More
Cupric Sulfate Pentahydrate (Fine Crystals/Certified ACS), Fisher Chemical™
$387.25 - $3515.25
Chemical Identifiers
CAS | 7758-99-8 |
---|---|
Molecular Formula | CuH10O9S |
Molecular Weight (g/mol) | 249.68 |
MDL Number | MFCD00149681 |
InChI Key | JZCCFEFSEZPSOG-UHFFFAOYSA-L |
Synonym | copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas |
PubChem CID | 24463 |
ChEBI | CHEBI:31440 |
IUPAC Name | copper(2+) pentahydrate sulfate |
SMILES | O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
C493-500
|
Thermo Fisher Scientific
FLC493500 |
500 g | Poly Bottle |
Each for $387.25
Case of 6 Each for $2,323.50
|
|
||||
C493-3
|
Fisher Chemical
FLC4933 |
3 kg | Poly Bottle |
Case of 4 Each for $2,885.83
|
|
||||
C493-10
|
Fisher Chemical
FLC49310 |
10 kg | Poly Pail |
Each for $3,515.25
|
|
||||
Chemical Identifiers
7758-99-8 | |
249.68 | |
JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
24463 | |
copper(2+) pentahydrate sulfate |
CuH10O9S | |
MFCD00149681 | |
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas | |
CHEBI:31440 | |
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
Specifications
150°C | |
7758-99-8 | |
3.5 to 4.5 | |
500 g | |
CuH10O9S | |
3077 | |
JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
copper(2+) pentahydrate sulfate | |
24463 | |
249.68 | |
Certified ACS | |
Poly Bottle | |
2.28g/mL |
110°C | |
Blue | |
Solid | |
98.0 to 102.0% | |
MFCD00149681 | |
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas | |
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O | |
249.68 | |
CHEBI:31440 | |
98.0 to 102.0% | |
Pass Test | |
7.3mmHg at 25°C | |
Cupric Sulfate Pentahydrate |
Safety and Handling
WARNING!
Emergency Overview
Harmful if swallowed. May cause irritation of respiratory tract. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.
NFPA
Health:2
Flammability:0
Instability:1
DOTInformation : DOT Class 9, : Miscellaneous