missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cupriethylenediamine, 1 M, Ricca Chemical

$477.56 - $1160.00

Chemical Identifiers

CAS 13426-91-0
Molecular Formula C4H12CuN4-2
Molecular Weight (g/mol) 179.714
InChI Key GOYYUYNOGNSLTE-UHFFFAOYSA-N
Synonym copper-ethylenediamine complex, cupriethylene diamine, bis 1,2-ethanediamine-n,n' copper 2+, koplex aquatic herbicide, unii-nip4i4lvcc, cupriethylenediamine, bis ethylenediamine copper ion, bis ethylenediamine copper 2+, bis ethylenediamine copper 2+ ion, ethane, 1,2-diamino-, copper complex
PubChem CID 4578819
IUPAC Name copper;2-azanidylethylazanide
SMILES C(C[NH-])[NH-].C(C[NH-])[NH-].[Cu+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
249932
View Documents
Ricca Chemical Company
249932
1 L Amber Glass Bottle
Each for $477.56
 
24991
SDS View Documents
Ricca Chemical Company
RI24991
4 L Amber Glass Bottle
Each for $1,160.00
 
Description

Description

Specifications

Chemical Identifiers

13426-91-0
179.714
copper-ethylenediamine complex, cupriethylene diamine, bis 1,2-ethanediamine-n,n' copper 2+, koplex aquatic herbicide, unii-nip4i4lvcc, cupriethylenediamine, bis ethylenediamine copper ion, bis ethylenediamine copper 2+, bis ethylenediamine copper 2+ ion, ethane, 1,2-diamino-, copper complex
copper;2-azanidylethylazanide
C4H12CuN4-2
GOYYUYNOGNSLTE-UHFFFAOYSA-N
4578819
C(C[NH-])[NH-].C(C[NH-])[NH-].[Cu+2]

Specifications

Blue to Purple
78.0,18.0
C4H12CuN4-2
GOYYUYNOGNSLTE-UHFFFAOYSA-N
copper;2-azanidylethylazanide
4578819
1 L
1Molar
13426-91-0,7732-18-5
82.0,22.0
copper-ethylenediamine complex, cupriethylene diamine, bis 1,2-ethanediamine-n,n' copper 2+, koplex aquatic herbicide, unii-nip4i4lvcc, cupriethylenediamine, bis ethylenediamine copper ion, bis ethylenediamine copper 2+, bis ethylenediamine copper 2+ ion, ethane, 1,2-diamino-, copper complex
C(C[NH-])[NH-].C(C[NH-])[NH-].[Cu+2]
179.714
Amber Glass Bottle
Liquid
Cupriethylenediamine
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'