missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 98+%

$129.76 - $384.30

Chemical Identifiers

CAS 458-37-7
Molecular Formula C21H20O6
Molecular Weight (g/mol) 368.39
MDL Number MFCD00008365
InChI Key ZIUSSTSXXLLKKK-KOBPDPAPSA-N
Synonym curcumin, diferuloylmethane, natural yellow 3, turmeric yellow, turmeric, curcuma, kacha haldi, gelbwurz, halad, curcumin i
PubChem CID 969516
ChEBI CHEBI:3962
SMILES COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC218580100
SDS View Documents
Thermo Scientific Chemicals
218580100
10 g Glass bottle
Each for $129.76
 
AC218580500
SDS View Documents
Thermo Scientific Chemicals
218580500
50 g Glass bottle
Each for $384.30
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

458-37-7
368.39
ZIUSSTSXXLLKKK-KOBPDPAPSA-N
969516
COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O
C21H20O6
MFCD00008365
curcumin, diferuloylmethane, natural yellow 3, turmeric yellow, turmeric, curcuma, kacha haldi, gelbwurz, halad, curcumin i
CHEBI:3962

Specifications

458-37-7
Orange
98+%
C21H20O6
[HOC6H3(OCH3)CH=CHCO]2CH2
10 g
15,2663
Solubility in water: insoluble. Other solubilities: insoluble in diethyl ether,soluble in alcohol,glacial acetic acid,acetone
COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O
368.39
CHEBI:3962
98+%
Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin)
183.0°C
Authentic
Glass bottle
1.4155 to 1.4175
MFCD00008365
08,IV,3697
curcumin, diferuloylmethane, natural yellow 3, turmeric yellow, turmeric, curcuma, kacha haldi, gelbwurz, halad, curcumin i
ZIUSSTSXXLLKKK-KOBPDPAPSA-N
(1E,6E)-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione
969516
368.38
Fine Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Warning

EINECSNumber : 207-280-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only