M+A Matting Cushion Station™ Anti-Fatigue Mats
Cushion Station™ Anti-Fatigue Mats feature nitrile-blended foam designed to withstand grease, oil, and chemicals. Anti-microbially treated to prevent odors.
$81.77 - $1275.67
Specifications
Certifications/Compliance NFSI Certified. Made in USA
Color Black
Thickness (English) 7/16 in.
View More Specs
Products 19
Catalog Number Mfr. No. Style Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Style Dimensions (L x W) Price Quantity  
17555078
View Documents
M+A Matting
3700000232
No holes 2 x 3.2 ft.
Each for $84.96
17555080
View Documents
M+A Matting
3700003283
No holes 3.2 x 8.3 ft.
Each for $418.66
17555081
View Documents
M+A Matting
3700032123
No holes 3.2 x 12.3 ft.
Each for $608.47
17555082
View Documents
M+A Matting
3700032161
No holes 3.2 x 16.1 ft.
Each for $814.04
17555083
View Documents
M+A Matting
3700032201
No holes 3.2 x 20.1 ft.
Each for $1,087.93
17555084
View Documents
M+A Matting
3700004592
No holes 4 x 5.92 ft.
Each for $357.95
17555085
View Documents
M+A Matting
3700000483
No holes 4 x 8.3 ft.
Each for $503.65
17555086
View Documents
M+A Matting
3700004123
No holes 4 x 12.3 ft.
Each for $760.62
17555088
View Documents
M+A Matting
3700004201
No holes 4 x 20.1 ft.
Each for $1,275.67
17555089
View Documents
M+A Matting
3710000232
With holes 2 x 3.2 ft.
Each for $81.77
17555090
View Documents
M+A Matting
3710003253
With holes 3.2 x 5.3 ft.
Each for $253.95
17555091
View Documents
M+A Matting
3710003283
With holes 3.2 x 8.3 ft.
Each for $402.91
17555092
View Documents
M+A Matting
3710032123
With holes 3.2 x 12.3 ft.
Each for $608.47
17555093
View Documents
M+A Matting
3710032161
With holes 3.2 x 16.1 ft.
Each for $814.04
17555094
View Documents
M+A Matting
3710032201
With holes 3.2 x 20.1 ft.
Each for $1,087.93
17555095
View Documents
M+A Matting
3710004592
With holes 4 x 5.92 ft.
Each for $357.95
17555096
View Documents
M+A Matting
3710000483
With holes 4 x 8.3 ft.
Each for $503.65
17555097
View Documents
M+A Matting
3710004123
With holes 4 x 12.3 ft.
Each for $760.62
17555099
View Documents
M+A Matting
3710004201
With holes 4 x 20.1 ft.
Each for $1,275.67
Description
Description
  • Comfortable - 7/16 in. closed-cell nitrile foam provides excellent anti-fatigue properties
  • Clean - Anti-microbial treatment provides lifetime protection from odors and degradation
  • Safe - Beveled edges provide a safe transition from floor to mat; certified high-traction by the National Floor Safety Institute (NFSI)
  • Versatile - Grease/oil proof, chemical resistant, welding safe
  • Easy to Clean - Flexible construction makes this mat easy to clean
  • Can be autoclave sterilized 
  • Available with or without holes for use in wet or dry areas
Specifications
Specifications
NFSI Certified. Made in USA
Black
7/16 in.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'