Reichert™ Cuvets for Reichert Unistat™ Bilirubinometer Edge
Edge
For use with Reichert UNISTAT Bilirubinometer (cat. no. AO10310C)
Supplier:  Reichert™ 13102140
 View more versions of this product
Catalog No. RE10214
Edge
Explore available promotions
Description
Description
  • For use with Unistat Bilirubinometer
  • Capacity: 20μL
Specifications
Specifications
Disposable Cuvette
Undiluted Serum
Reichert Unistat™ Bilirubinometer
20 μL