missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cyanide Standard, 1000 ppm CN-, Ricca Chemical
Supplier: Ricca Chemical Company 254332
Specifications
Cyanide Standard | |
Amber Poly Bottle | |
Colorless | |
CKN | |
[C-]#N.[K+] | |
65.116 | |
1 mL = 1 mg CN |
151-50-8,1310-73-2,7732-18-5 | |
1 L | |
1000ppm CN | |
NNFCIKHAZHQZJG-UHFFFAOYSA-N | |
potassium;cyanide | |
9032 | |
Liquid |
Chemical Identifiers
151-50-8 | |
65.116 | |
9032 | |
[C-]#N.[K+] |
CKN | |
NNFCIKHAZHQZJG-UHFFFAOYSA-N | |
potassium;cyanide |