missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyanuric acid, 99%, Thermo Scientific Chemicals

$79.21 - $225.92

Chemical Identifiers

CAS 108-80-5
Molecular Formula C3H3N3O3
Molecular Weight (g/mol) 129.08
MDL Number MFCD00082990
InChI Key ZFSLODLOARCGLH-UHFFFAOYSA-N
Synonym cyanuric acid, isocyanuric acid, 1,3,5-triazine-2,4,6-triol, trihydroxycyanidine, tricyanic acid, sym-triazinetriol, s-triazinetriol, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, pseudocyanuric acid, isocyanurate acid
PubChem CID 7956
ChEBI CHEBI:38028
IUPAC Name 1,3,5-triazinane-2,4,6-trione
SMILES O=C1NC(=O)NC(=O)N1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1544722
View Documents
Thermo Scientific Chemicals
A1544722
100 g
Each for $79.21
 
AAA1544730
View Documents
Thermo Scientific Chemicals
A1544730
250 g
Each for $114.74
 
AAA154470B
View Documents
Thermo Scientific Chemicals
A154470B
1000 g
Each for $225.92
 
Description

Description

Cyanuric acid acts as a chlorine stabilizer in swimming pools. It is used as a precursor to N-chlorinated cyanurates, which are used to disinfect water and crosslinking agents for polyurethane resins. It serves as a process regulator. Further, it is used in the diagnostic determination of melamine and related compounds in kidney tissue.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in sulfuric acid, dimethylformamide, sodium hydroxide, potassium hydroxide and diemthyl sulfoxide. Slightly soluble in acetone, benzene, diethyl ether, ethanol and hexane. Insoluble in cold methanol, benzene and chloroform.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

108-80-5
129.08
ZFSLODLOARCGLH-UHFFFAOYSA-N
7956
1,3,5-triazinane-2,4,6-trione
C3H3N3O3
MFCD00082990
cyanuric acid, isocyanuric acid, 1,3,5-triazine-2,4,6-triol, trihydroxycyanidine, tricyanic acid, sym-triazinetriol, s-triazinetriol, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, pseudocyanuric acid, isocyanurate acid
CHEBI:38028
O=C1NC(=O)NC(=O)N1

Specifications

>360°C
99%
MFCD00082990
14,2698
Soluble in sulfuric acid,dimethylformamide,sodium hydroxide,potassium hydroxide and diemthyl sulfoxide. Slightly soluble in acetone,benzene,diethyl ether,ethanol and hexane. Insoluble in cold methanol,benzene and chloroform.
O=C1NC(=O)NC(=O)N1
129.08
CHEBI:38028
99%
100 g
108-80-5
C3H3N3O3
126982
cyanuric acid, isocyanuric acid, 1,3,5-triazine-2,4,6-triol, trihydroxycyanidine, tricyanic acid, sym-triazinetriol, s-triazinetriol, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, pseudocyanuric acid, isocyanurate acid
ZFSLODLOARCGLH-UHFFFAOYSA-N
1,3,5-triazinane-2,4,6-trione
7956
129.08
2.5
Cyanuric acid
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 203-618-0

RTECSNumber : XZ1800000

TSCA : Yes

storageNote1 : Hygroscopic

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only