Learn More
Cyclobutanol, 96%, Thermo Scientific Chemicals
$220.51 - $2079.66
Chemical Identifiers
CAS | 2919-23-5 |
---|---|
Molecular Formula | C4H8O |
Molecular Weight (g/mol) | 72.11 |
MDL Number | MFCD00001318 |
InChI Key | KTHXBEHDVMTNOH-UHFFFAOYSA-N |
Synonym | cyclobutyl alcohol, cyclobutyl hydroxide, hydroxycyclobutane, acmc-1cgim, cyclobutanol, cyclobutanol 1g, ksc206s6t, kthxbehdvmtnoh-uhfffaoysa |
PubChem CID | 76218 |
IUPAC Name | cyclobutanol |
SMILES | OC1CCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1243003
|
Thermo Scientific Chemicals
A1243003 |
1 g |
Each for $220.51
|
|
|||||
AAA1243006
|
Thermo Scientific Chemicals
A1243006 |
5 g |
Each for $675.44
|
|
|||||
AAA1243014
|
Thermo Scientific Chemicals
A1243014 |
25 g |
Each for $2,079.66
|
|
|||||
Description
Has synthetic applications of gold-catalyzed ring expansions. Development of novel type of transition metal-catalyzed cascade reactions utilizing small ring systems and the applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHas synthetic applications of gold-catalyzed ring expansions. Development of novel type of transition metal-catalyzed cascade reactions utilizing small ring systems and the applications.
Solubility
Partly soluble.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid chlorides, acid anhydrides.
Chemical Identifiers
2919-23-5 | |
72.11 | |
KTHXBEHDVMTNOH-UHFFFAOYSA-N | |
76218 | |
OC1CCC1 |
C4H8O | |
MFCD00001318 | |
cyclobutyl alcohol, cyclobutyl hydroxide, hydroxycyclobutane, acmc-1cgim, cyclobutanol, cyclobutanol 1g, ksc206s6t, kthxbehdvmtnoh-uhfffaoysa | |
cyclobutanol |
Specifications
2919-23-5 | |
123°C to 124°C | |
C4H8O | |
MFCD00001318 | |
UN1120 | |
cyclobutyl alcohol, cyclobutyl hydroxide, hydroxycyclobutane, acmc-1cgim, cyclobutanol, cyclobutanol 1g, ksc206s6t, kthxbehdvmtnoh-uhfffaoysa | |
KTHXBEHDVMTNOH-UHFFFAOYSA-N | |
cyclobutanol | |
76218 | |
96% |
0.925 | |
21°C (69°F) | |
1.436 | |
1 g | |
2035937 | |
Partly soluble. | |
OC1CCC1 | |
72.11 | |
72.11 | |
Cyclobutanol |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTANOLS
EINECSNumber : 220-858-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only