missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cyclobutanone (stabilized with Na2CO3) 97.0+%, TCI America™
$91.24 - $298.62
Chemical Identifiers
CAS | 1191-95-3 |
---|---|
Molecular Formula | C4H6O |
Molecular Weight (g/mol) | 70.09 |
MDL Number | MFCD00001332 |
InChI Key | SHQSVMDWKBRBGB-UHFFFAOYSA-N |
Synonym | unii-6pf2sh405u, cyclobutyloxy, cyclobutanone, 98+%, cyclobutanon, cylcobutanone, cylobutanone, 3-cyclobutanone, 1-oxocyclobutane, cyclobutanone, pubchem9021 |
PubChem CID | 14496 |
IUPAC Name | cyclobutanone |
SMILES | O=C1CCC1 |
Chemical Identifiers
1191-95-3 | |
70.09 | |
SHQSVMDWKBRBGB-UHFFFAOYSA-N | |
14496 | |
O=C1CCC1 |
C4H6O | |
MFCD00001332 | |
unii-6pf2sh405u, cyclobutyloxy, cyclobutanone, 98+%, cyclobutanon, cylcobutanone, cylobutanone, 3-cyclobutanone, 1-oxocyclobutane, cyclobutanone, pubchem9021 | |
cyclobutanone |
Specifications
1191-95-3 | |
Yellow | |
C4H6O | |
1 g | |
unii-6pf2sh405u, cyclobutyloxy, cyclobutanone, 98+%, cyclobutanon, cylcobutanone, cylobutanone, 3-cyclobutanone, 1-oxocyclobutane, cyclobutanone, pubchem9021 | |
O=C1CCC1 | |
70.09 | |
70.09 | |
Liquid |
-53°C | |
99°C | |
MFCD00001332 | |
1224 | |
SHQSVMDWKBRBGB-UHFFFAOYSA-N | |
cyclobutanone | |
14496 | |
≥97.0% (GC) | |
Cyclobutanone (stabilized with Na2CO3) |
Safety and Handling
TSCA : Yes
Recommended Storage : Freezer