Learn More
Cyclobutyl bromide, 97%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 320500010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cyclobutyl bromide | |
Colorless | |
108.0°C | |
Authentic | |
Glass bottle | |
1.4776 to 1.4797 | |
MFCD00001317 | |
1.43 | |
Solubility in water: immiscible | |
C1CC(C1)Br | |
135.01 | |
135.01 | |
Pure |
4399-47-7 | |
1.4300g/mL | |
22°C | |
96% min. (GC) | |
C4H7Br | |
C4H7Br | |
1 g | |
cyclobutyl bromide, cyclobutane, bromo, cyclobutylbromid, cyclobutylbromide, bromo cyclobutane, bromo-cyclobutane, bromocyclobutane, acmc-209jw1, ksc235o4p, inchi=1/c4h7br/c5-4-2-1-3-4/h4h,1-3h | |
KXVUSQIDCZRUKF-UHFFFAOYSA-N | |
bromocyclobutane | |
78110 | |
97% | |
Liquid |
Chemical Identifiers
4399-47-7 | |
135.01 | |
KXVUSQIDCZRUKF-UHFFFAOYSA-N | |
78110 | |
C1CC(C1)Br |
C4H7Br | |
MFCD00001317 | |
cyclobutyl bromide, cyclobutane, bromo, cyclobutylbromid, cyclobutylbromide, bromo cyclobutane, bromo-cyclobutane, bromocyclobutane, acmc-209jw1, ksc235o4p, inchi=1/c4h7br/c5-4-2-1-3-4/h4h,1-3h | |
bromocyclobutane |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a g
GHS Signal Word: Danger
EINECSNumber : 224-530-9
RUO – Research Use Only