Learn More
Cyclobutylamine, 98%
CAS: 2516-34-9 | C4H9N | 71.123 g/mol
$104.32 - $1567.59
Chemical Identifiers
| CAS | 2516-34-9 |
|---|---|
| Molecular Formula | C4H9N |
| Molecular Weight (g/mol) | 71.123 |
| MDL Number | MFCD00001328 |
| InChI Key | KZZKOVLJUKWSKX-UHFFFAOYSA-N |
| Synonym | cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride |
| PubChem CID | 75645 |
| IUPAC Name | cyclobutanamine |
| SMILES | C1CC(C1)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1342303
|
Thermo Scientific Chemicals
A1342303 |
1 g |
Each for $104.32
|
|
|||||
|
AAA1342306
|
Thermo Scientific Chemicals
A1342306 |
5 g |
Each for $344.23
|
|
|||||
|
AAA1342314
|
Thermo Scientific Chemicals
A1342314 |
25 g |
Each for $1,567.59
|
|
|||||
Description
Substrate employed in a microwave-assisted synthesis of 7-azaindoles from dihalopyridines. Also it is a reactant in the preparation of imidazo[1,2-b]pyridazine derivatives as selective and orally available Mps1 (TTK) kinase inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSubstrate employed in a microwave-assisted synthesis of 7-azaindoles from dihalopyridines. Also it is a reactant in the preparation of imidazo[1,2-b]pyridazine derivatives as selective and orally available Mps1 (TTK) kinase inhibitors.
Solubility
Slightly soluble in water.
Notes
Hygroscopic, store way form water/moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agents.
Chemical Identifiers
| 2516-34-9 | |
| 71.123 | |
| KZZKOVLJUKWSKX-UHFFFAOYSA-N | |
| 75645 | |
| C1CC(C1)N |
| C4H9N | |
| MFCD00001328 | |
| cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride | |
| cyclobutanamine |
Specifications
| 2516-34-9 | |
| 82°C to 83°C | |
| C4H9N | |
| MFCD00001328 | |
| UN2733 | |
| Air sensitive; Hygroscopic | |
| Slightly soluble in water. | |
| C1CC(C1)N | |
| 71.123 | |
| 71.12 | |
| Cyclobutylamine |
| 0.836 | |
| −4°C (24°F) | |
| 1.4376 | |
| 1 g | |
| 2069297 | |
| cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride | |
| KZZKOVLJUKWSKX-UHFFFAOYSA-N | |
| cyclobutanamine | |
| 75645 | |
| 98% |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 219-736-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only