missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclocytidine hydrochloride, 98+%, Thermo Scientific Chemicals

Anti-tumor agent

Supplier:  Thermo Scientific Chemicals J6384503

 View more versions of this product

Catalog No. AAJ6384503


Explore available promotions

Description

Description

Anti-tumor agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclocytidine hydrochloride
248°C to 250°C
C9H12ClN3O4
1 g
cyclocytidine hydrochloride 1g
Cl.OC[C@H]1O[C@@H]2[C@@H](OC3=NC(=N)C=CN23)[C@@H]1O
261.66
CHEBI:74843
≥98%
10212-25-6
White
MFCD00012636
14,629
KZOWNALBTMILAP-JBMRGDGGSA-N
(2R,3R,3aS,9aR)-2-(hydroxymethyl)-6-imino-2,3,3a,9a-tetrahydrofuro[1,2][1,3]oxazolo[3,4-a]pyrimidin-3-ol;hydron;chloride
74764394
261.66
Powder

Chemical Identifiers

10212-25-6
261.66
KZOWNALBTMILAP-JBMRGDGGSA-N
74764394
(2R,3R,3aS,9aR)-2-(hydroxymethyl)-6-imino-2,3,3a,9a-tetrahydrofuro[1,2][1,3]oxazolo[3,4-a]pyrimidin-3-ol;hydron;chloride
C9H12ClN3O4
MFCD00012636
cyclocytidine hydrochloride 1g
CHEBI:74843
Cl.OC[C@H]1O[C@@H]2[C@@H](OC3=NC(=N)C=CN23)[C@@H]1O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 233-515-6

RTECSNumber : LV2615000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only