missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
β-Cyclodextrin, 98%, Thermo Scientific Chemicals
$73.75 - $401.37
Chemical Identifiers
CAS | 7585-39-9 |
---|---|
Molecular Formula | C42H70O35 |
Molecular Weight (g/mol) | 1134.99 |
MDL Number | MFCD00078139 |
InChI Key | WHGYBXFWUBPSRW-UHFFFAOYNA-N |
Synonym | cycloheptaamylose, cycloheptapentylose, .beta.-cyclodextrin, caraway, cyclo-hepta-amylose, cycloheptamaltose, .beta.-dextrin, beta-cycloamylose, beta-cyclodextrin, .beta.-cycloamylose |
PubChem CID | 131707246 |
IUPAC Name | 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,â¶.2â¸,¹¹.2¹³,¹â¶.2¹â¸,²¹.2²³,²â¶.2²â¸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol |
SMILES | OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC406000250
|
Thermo Scientific Chemicals
406000250 |
25 g |
Each for $73.75
|
|
|||||
AC406001000
|
Thermo Scientific Chemicals
406001000 |
100 g |
Each for $156.56
|
|
|||||
AC406005000
|
Thermo Scientific Chemicals
406005000 |
500 g |
Each for $401.37
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7585-39-9 | |
1134.99 | |
WHGYBXFWUBPSRW-UHFFFAOYNA-N | |
131707246 | |
OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O |
C42H70O35 | |
MFCD00078139 | |
cycloheptaamylose, cycloheptapentylose, .beta.-cyclodextrin, caraway, cyclo-hepta-amylose, cycloheptamaltose, .beta.-dextrin, beta-cycloamylose, beta-cyclodextrin, .beta.-cycloamylose | |
5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,â¶.2â¸,¹¹.2¹³,¹â¶.2¹â¸,²¹.2²³,²â¶.2²â¸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol |
Specifications
7585-39-9 | |
14% max. (105°C, 8 hrs) | |
97.5% min. (ELSD) (HPLC) | |
MFCD00078139 | |
06,151; 08,133; 09,129; 12,151; 15,107 | |
+ 150.00 (25.00°C c=1.5, in water) | |
cycloheptaamylose, cycloheptapentylose, .beta.-cyclodextrin, caraway, cyclo-hepta-amylose, cycloheptamaltose, .beta.-dextrin, beta-cycloamylose, beta-cyclodextrin, .beta.-cycloamylose | |
WHGYBXFWUBPSRW-UHFFFAOYNA-N | |
5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,â¶.2â¸,¹¹.2¹³,¹â¶.2¹â¸,²¹.2²³,²â¶.2²â¸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol | |
131707246 | |
98% |
310.0°C | |
Authentic | |
C42H70O35 | |
25 g | |
15,271 | |
+ 150.00 | |
Solubility in water: soluble. | |
OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O | |
1134.99 | |
1134.98 | |
β-Cyclodextrin |
Safety and Handling
EINECSNumber : 231-493-2
RUO – Research Use Only