missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cycloheptane, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 111010050

Catalog No. AC111010050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Cycloheptane
0.8100g/mL
6°C
98.5% min. (GC)
C7H14
C7H14
5mL
0.81
Solubility in water: insoluble. Other solubilities: soluble in alcohol
C1CCCCCC1
98.19
98.19
291-64-5
118.4°C
Authentic
Glass bottle
1.4440 to 1.446
MFCD00004147
05,29
heptamethylene, unii-vtz53p34ja, vtz53p34ja, cycloheptan, hsdb 59, cycloheptane, un2241 flammable liquid
DMEGYFMYUHOHGS-UHFFFAOYSA-N
cycloheptane
9265
99%
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-030-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only